ADV465748878 AChemBlock

(3-Methyl-piperidin-4-yl)-acetic acid ethyl ester hydrochloride

  • MDL number MFCD28166358
  • Cas Number : 1630907-26-4
  • Molecular Weight : 221.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White powder
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-piperidin-4-yl-acetic-acid-ethyl-ester-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9863
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart