ADV465748890 AChemBlock

4-Aminomethyl indazole dihydrochloride

  • MDL number MFCD28166362
  • Cas Number : 1630907-20-8
  • Molecular Weight : 220.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-aminomethyl-indazole-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9688
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart