ADV465748893 AChemBlock

Ethyl 3-chloro-1-methyl-1H-pyrazole-5-carboxylate

  • MDL number MFCD19704132
  • Cas Number : 173841-07-1
  • Molecular Weight : 188.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3-chloro-1-methyl-1h-pyrazole-5-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9775
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart