ADV465748897 AChemBlock

1-Boc-3-amino-4-(hydroxymethyl)pyrrolidine

  • MDL number MFCD11847979
  • Cas Number : 1184918-34-0
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-amino-4-hydroxymethyl-pyrrolidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10003
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart