ADV465748901 AChemBlock

tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoindoline-2-carboxylate

  • MDL number MFCD11226842
  • Cas Number : 905273-91-8
  • Molecular Weight : 345.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2h-isoindole-2-carboxylicacid-1-3-dihydro-5-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-1-1-dimethylethyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10008
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart