ADV465748904 AChemBlock

cis-tert-Butyl octahydro-1H-pyrrolo[3,4-b]pyridine-1-carboxylate

  • MDL number MFCD12198664
  • Cas Number : 181141-40-2
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/cis-tert-butyl-octahydro-1h-pyrrolo-3-4-b-pyridine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10011
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart