ADV465748906 AChemBlock

5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylic acid

  • MDL number MFCD11846936
  • Cas Number : 796729-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-6-dihydro-4h-pyrrolo-1-2-b-pyrazole-2-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10013
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart