ADV465748913 AChemBlock

(1S,3S)-N-Boc-3-aminocyclopentanecarboxylic acid

  • MDL number MFCD02259726
  • Cas Number : 161601-29-2
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1s-3s-n-boc-3-aminocyclopentanecarboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10020
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart