ADV465748916 AChemBlock

2-Oxa-8-azaspiro[4.5]decane

  • MDL number MFCD11035890
  • Cas Number : 176-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-oxa-8-azaspiro-4-5-decane.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10023
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart