ADV465748919 AChemBlock

(R)-4-(Hydroxymethyl)oxazolidin-2-one

  • MDL number MFCD11847629
  • Cas Number : 132682-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/r-4-hydroxymethyl-oxazolidin-2-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10026
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart