ADV465748922 AChemBlock

cis-1-Boc-3-hydroxy-4-aminomethylpyrrolidine

  • MDL number MFCD20126210
  • Cas Number : 872714-78-8
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/cis-1-boc-3-hydroxy-4-aminomethylpyrrolidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10029
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart