ADV465748934 AChemBlock

(S)-2,3-Dihydro-1,4-benzodioxin-2-methanamine

  • MDL number MFCD10697676
  • Cas Number : 46049-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-2-3-dihydro-1-4-benzodioxin-2-methanamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10041
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart