ADV465748937 AChemBlock

4-((R)-hydroxy((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methyl)quinolin-1-ium (1R,4R,5R)-3-oxo-2-oxabicyclo[2.2.1]heptane-5-carboxylate

  • MDL number MFCD28023827
  • Cas Number : 862174-60-5
  • Molecular Weight : 450.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-oxabicyclo-2-2-1-heptane-5-carboxylic-acid-3-oxo-1r-4r-5r-cinchonidine-salt.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5890
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart