ADV465748947 AChemBlock

cis-3-[(tert-Butoxycarbonyl)amino]cyclopentane-1-carboxylic acid

  • MDL number MFCD09992879
  • Cas Number : 410090-37-8
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/cis-3-tert-butoxycarbonyl-amino-cyclopentane-1-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10054
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart