ADV465748950 AChemBlock

methyl (S)-3-((tert-butoxycarbonyl)amino)-4-hydroxybutanoate

  • MDL number MFCD28023842
  • Cas Number : 136703-59-8
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/butanoic-acid-3-1-1-dimethylethoxy-carbonyl-amino-4-hydroxy-methyl-ester-3s.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10057
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart