ADV465748957 AChemBlock

(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid

  • MDL number MFCD00671385
  • Cas Number : 109183-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-2-tert-butoxycarbonyl-amino-2-cyclopentylacetic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10064
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart