ADV465749013 AChemBlock

Potassium (morpholin-4-yl)methyltrifluoroborate

  • MDL number MFCD10566512
  • Cas Number : 936329-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/potassium-morpholin-4-yl-methyltrifluoroborate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10124
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart