ADV465749023 AChemBlock

tert-Butyl spiro[indoline-3,4'-piperidine]-1-carboxylate

  • MDL number MFCD09266212
  • Cas Number : 676607-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-spiro-indoline-3-4-piperidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10134
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart