ADV465749054 AChemBlock

rel-methyl (1R,3R)-3-((tert-butoxycarbonyl)amino)cyclopentane-1-carboxylate

  • MDL number MFCD27977593
  • Cas Number : 862700-37-6
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/cyclopentanecarboxylic-acid-3-1-1-dimethylethoxy-carbonyl-amino-methyl-ester-1r-3r-rel.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10165
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart