ADV465749077 AChemBlock

Methyl 5,5-dimethyl-2-oxocyclohexanecarboxylate

  • MDL number MFCD09869052
  • Cas Number : 50388-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-5-dimethyl-2-oxocyclohexanecarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10188
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart