ADV465749090 AChemBlock

N-Boc-2-morpholinecarbaldehyde

  • MDL number MFCD06410632
  • Cas Number : 218594-02-6
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/n-boc-2-morpholinecarbaldehyde.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10201
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart