ADV465749115 AChemBlock

1-(tert-Butyl) 4-ethyl 5-(4-fluorophenyl)-3,6-dihydropyridine-1,4(2H)-dicarboxylate

  • MDL number MFCD29037373
  • Cas Number : 1432675-99-4
  • Molecular Weight : 351.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-tert-butyl-4-ethyl-5-4-fluorophenyl-3-6-dihydropyridine-1-4-2h-dicarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8678
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart