ADV465749173 AChemBlock

AUY922

  • MDL number MFCD14635361
  • Cas Number : 747412-49-3
  • Molecular Weight : 465.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/auy922.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10274
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart