ADV465749198 AChemBlock

CGI-1746

  • MDL number MFCD18782602
  • Cas Number : 910232-84-7
  • Molecular Weight : 579.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/cgi-1746.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10301
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart