ADV465749242 AChemBlock

LY2157299

  • MDL number MFCD12923319
  • Cas Number : 700874-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ly2157299.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10348
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart