ADV465749265 AChemBlock

Salubrinal

  • MDL number MFCD00548612
  • Cas Number : 405060-95-9
  • Molecular Weight : 479.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/salubrinal.html

Safety Information

signalword Warning
pcodes P261;P264;P271;P280;P302+352;P304+340;P305+351+338;P312;P321;P332+313;P337+313;P362;P403+235;P405;P501
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10374
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart