ADV465749365 AChemBlock

Teniposide

  • MDL number MFCD00866516
  • Cas Number : 29767-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/teniposide.html

Safety Information

signalword Danger
pcodes P201;P202;P281;P308+313;P405;P501
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10486
pictograms GHS08
hstatements H350
Availability details for
Quantity
  • Availability
  • Add to Cart