ADV465749450 AChemBlock

5-Trifluoromethyl-4-azaindole

  • MDL number MFCD12962601
  • Cas Number : 1190315-94-6
  • Molecular Weight : 186.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-trifluoromethyl-4-azaindole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10577
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart