ADV465749485 AChemBlock

Ethyl 5-bromo-1H-1,2,4-triazole-3-carboxylate

  • MDL number MFCD11040185
  • Cas Number : 774608-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ethyl-5-bromo-1h-1-2-4-triazole-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10612
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart