ADV465749490 AChemBlock

1,3-Dihydro-3,3-dimethyl-2H-indol-2-one

  • MDL number MFCD00158549
  • Cas Number : 19155-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-3-dihydro-3-3-dimethyl-2h-indol-2-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10617
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart