ADV465749495 AChemBlock

(+)-(3R,4R)-3,4-dimethyl-4-(3-hydroxyphenyl)piperidine

  • MDL number MFCD06659960
  • Cas Number : 119193-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3r-4r-3-4-dimethyl-4-3-hydroxyphenyl-piperidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10622
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart