ADV465749496 AChemBlock

N-(1H-benzimidazol-2-ylmethyl)-N-[(8S)-5,6,7,8-tetrahydro-8-quinolinyl]- (9CI)

  • MDL number MFCD11977316
  • Cas Number : 558447-26-0
  • Molecular Weight : 349.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/n-1h-benzimidazol-2-ylmethyl-n-8s-5-6-7-8-tetrahydro-8-quinolinyl-9ci.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10623
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart