ADV465749500 AChemBlock

10-Propargyl-10-deazaaminopterin

  • MDL number MFCD00920897
  • Cas Number : 146464-95-1
  • Molecular Weight : 477.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/10-propargyl-10-deazaaminopterin.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10627
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart