ADV465749510 AChemBlock

5,7-Difluoroquinazolin-4(3H)-one

  • MDL number MFCD11100783
  • Cas Number : 379228-58-7
  • Molecular Weight : 182.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-7-difluoroquinazolin-4-3h-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10637
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart