ADV465749517 AChemBlock

4,5,6,7-Tetrahydrothiazolo[5,4-c]pyridine hydrochloride

  • MDL number MFCD08726044
  • Cas Number : 1241725-84-7
  • Molecular Weight : 176.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-5-6-7-tetrahydrothiazolo-5-4-c-pyridine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10644
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart