ADV465749543 AChemBlock

tert-butyl ((3R,4R)-3-hydroxypiperidin-4-yl)carbamate

  • MDL number MFCD18914363
  • Cas Number : 724788-29-8
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/carbamic-acid-3r-4r-3-hydroxy-4-piperidinyl-1-1-dimethylethyl-ester-9ci.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10671
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart