ADV465749550 AChemBlock

(3S,4S)-3-Amino-4-methyl-2-azetidinone hydrochloride

  • MDL number MFCD28134613
  • Cas Number : 1844898-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3s-4s-3-amino-4-methyl-2-azetidinone-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10678
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart