ADV465749552 AChemBlock

(R)-1-(t-Butoxycarbonyl)-2,3-oxiranylamine

  • MDL number MFCD11111972
  • Cas Number : 149057-20-5
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/r-1-t-butoxycarbonyl-2-3-oxiranylamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10680
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart