ADV465749558 AChemBlock

Methyl tetrahydro-4H-pyran-4-one-3-carboxylate

  • MDL number MFCD20921984
  • Cas Number : 127956-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-tetrahydro-4h-pyran-4-one-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10686
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart