ADV465749572 AChemBlock

(1R,2R)-2-(Methoxycarbonyl)cyclohexanecarboxylic acid

  • MDL number MFCD18071078
  • Cas Number : 96894-64-3
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1r-2r-2-methoxycarbonyl-cyclohexanecarboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10700
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart