ADV465749578 AChemBlock

3-Methylfuran

  • MDL number MFCD00060134
  • Cas Number : 930-27-8
  • Molecular Weight : 82.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-methylfuran.html

Safety Information

signalword Danger
pcodes P201;P202;P210;P233;P240;P301+330+331;P303+361+353
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10706
pictograms GHS02;GHS05;GHS06;GHS08;GHS09
hstatements H222;H302;H315;H331;H341;H351;H373;H411
Availability details for
Quantity
  • Availability
  • Add to Cart