ADV465749602 AChemBlock

Medetomidine HCl

  • MDL number MFCD06798319
  • Cas Number : 86347-15-1
  • Molecular Weight : 236.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/medetomidine-hcl.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=10731
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart