ADV465749677 AChemBlock

5-Hydroxy-1,2,3,3-tetramethyl-3H-indolium iodide

  • MDL number MFCD28166373
  • Cas Number : 59223-23-3
  • Molecular Weight : 317.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-hydroxy-1-2-3-3-tetramethyl-3h-indolium-iodide.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-987
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart