ADV465749678 AChemBlock

2,3-Dihydroxy-5-nitrobenzaldehyde

  • MDL number MFCD25964229
  • Cas Number : 52924-54-6
  • Molecular Weight : 183.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-3-dihydroxy-5-nitrobenzaldehyde.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-988
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart