ADV465749684 AChemBlock

3-(Trifluoromethyl)oxetane-3-amine hydrochloride

  • MDL number MFCD28155084
  • Cas Number : 1268883-21-1
  • Molecular Weight : 177.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-trifluoromethyl-oxetane-3-amine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8450-1
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart