ADV465749690 AChemBlock

3-Methylsulfanyl-piperidine hydrochloride

  • MDL number MFCD16039580
  • Cas Number : 1229627-46-6
  • Molecular Weight : 167.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-methylsulfanyl-piperidine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8554
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart