ADV465749693 AChemBlock

1-(Oxan-4-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD19611565
  • Cas Number : 1340372-11-3
  • Molecular Weight : 196.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-oxan-4-yl-1h-pyrazole-4-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8568
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart