ADV465749722 AChemBlock

1-(Tert-butoxycarbonyl)-3-(1-piperazinyl)azetidine

  • MDL number MFCD09264407
  • Cas Number : 178311-48-3
  • Molecular Weight : 241.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-tert-butoxycarbonyl-3-1-piperazinyl-azetidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8639
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart