ADV465749732 AChemBlock

tert-Butyl cis-4-amino-2-methyl-1-piperidinecarboxylate

  • MDL number MFCD24850074
  • Cas Number : 1434073-24-1
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-cis-4-amino-2-methyl-1-piperidinecarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8670
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart