ADV465749737 AChemBlock

(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propanoic acid

  • MDL number MFCD03412130
  • Cas Number : 216439-76-8
  • Molecular Weight : 391.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-2-tert-butoxycarbonyl-amino-3-4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-phenyl-propanoic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8687
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart